2-Hydroxy-2-(4-propoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A493098-10G A493098-1G
76877-972EA 43.5 USD
76877-972 76877-974
2-Hydroxy-2-(4-propoxyphenyl)acetic acid 97%
2-Hydroxy-2-(4-propoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
MDL Number: MFCD07368282
CAS Number: 79694-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR