4-Chloro-2-fluorophenylacetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-2-fluorobenzeneacetonitrile

A492208-1G A492208-25G A492208-5G
76877-966EA 5.31 USD
76877-966 76877-968 76877-970
4-Chloro-2-fluorophenylacetonitrile 98%
4-Chloro-2-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.59 g/mol
MDL Number: MFCD01312758
CAS Number: 75279-53-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR