N-Methyl-3-nitrobenzamide 98%

Supplier: Ambeed
Warning

A489570-1G A489570-250MG A489570-5G
76877-950EA 66.84 USD
76877-950 76877-952 76877-954
N-Methyl-3-nitrobenzamide 98%
N-Methyl-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₈H₈N₂O₃
MW: 180.161 g/mol
MDL Number: MFCD00714941
CAS Number: 3400-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR