4-(1-(Hydroxymethyl)cyclopropyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A487487-100MG A487487-1G A487487-250MG
76877-936EA 60 USD
76877-936 76877-938 76877-940
4-(1-(Hydroxymethyl)cyclopropyl)phenylboronic acid 98%
4-(1-(Hydroxymethyl)cyclopropyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MDL Number: MFCD12546571
CAS Number: 1217501-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR