4-(Methylcarbamoyl)phenylboronic acid propanediol cyclic ester 96%

Supplier: Ambeed
Warning

Synonyms: 4-(1,3,2-Dioxaborinan-2-yl)-N-methylbenzamide

A486835-1G A486835-250MG A486835-5G
76877-924EA 59 USD
76877-924 76877-926 76877-928
4-(Methylcarbamoyl)phenylboronic acid propanediol cyclic ester 96%
4-(Methylcarbamoyl)phenylboronic acid propanediol cyclic ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₃
MDL Number: MFCD09801045
CAS Number: 957120-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR