4-(3-Chloropropylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((3-Chloropropyl)carbamoyl)phenyl)boronic acid

A486367-1G A486367-250MG A486367-5G
76877-918EA 87 USD
76877-918 76877-920 76877-922
4-(3-Chloropropylcarbamoyl)phenylboronic acid 98%
4-(3-Chloropropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
MW: 241.4800 g/mol
MDL Number: MFCD08235077
CAS Number: 874460-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR