5-Chloro-(2-methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Chloro-2-(methoxycarbonyl)phenyl)boronic acid

A485041-1G A485041-250MG A485041-5G
76877-912EA 51 USD
76877-912 76877-914 76877-916
5-Chloro-(2-methoxycarbonyl)phenylboronic acid 98%
5-Chloro-(2-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₄
MW: 214.4100 g/mol
MDL Number: MFCD09878336
CAS Number: 957062-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR