1-Chloro-2-fluoro-5-iodo-3-(trifluoromethyl)benzene ≥98%

Supplier: Ambeed
Warning

A484430-1G A484430-250MG A484430-5G
76877-898EA 55 USD
76877-898 76877-900 76877-902
1-Chloro-2-fluoro-5-iodo-3-(trifluoromethyl)benzene ≥98%
1-Chloro-2-fluoro-5-iodo-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂ClF₄I
MW: 324.44 g/mol
MDL Number: MFCD22421634
CAS Number: 928783-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR