N-Phenylnicotinamide hydrochloride 95+%

Supplier: Ambeed
Warning

Synonyms: Nicotinanilide Hydrochloride

A484321-1G A484321-250MG A484321-5G
76877-892EA 28 USD
76877-892 76877-894 76877-896
N-Phenylnicotinamide hydrochloride 95+%
N-Phenylnicotinamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MDL Number: MFCD00060194
CAS Number: 69135-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR