(S)-3-Aminoazepan-2-one hydrochloride 97%

Supplier: Ambeed
Warning

A484172-10G A484172-1G A484172-5G
76877-886EA 76.39 USD
76877-886 76877-888 76877-890
(S)-3-Aminoazepan-2-one hydrochloride 97%
(S)-3-Aminoazepan-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O
MW: 164.63 g/mol
MDL Number: MFCD02683418
CAS Number: 26081-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR