Boc-Glu(Osu)-Otbu 95%

Supplier: Ambeed
Warning

A484044-1G A484044-5G
76877-882EA 68.99 USD
76877-882 76877-884
Boc-Glu(Osu)-Otbu 95%
Boc-Glu(Osu)-Otbu

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂O₈
MDL Number: MFCD00236848
CAS Number: 81659-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR