Moxonidine 98%

Supplier: Ambeed
Warning

A482917-100MG A482917-10MG A482917-1G A482917-250MG A482917-5G A482917-5MG A482917-1MG
76877-864EA 28.99 USD
76877-864 76877-866 76877-868 76877-870 76877-872 76877-874 77673-254
Moxonidine 98%
Moxonidine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₅O
MDL Number: MFCD00865794
CAS Number: 75438-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR