3-Iodo-6-methylpyridazine 95%

Supplier: Ambeed
Warning

A480899-1G A480899-250MG A480899-5G
76877-838EA 34 USD
76877-838 76877-840 77594-500
3-Iodo-6-methylpyridazine 95%
3-Iodo-6-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
MDL Number: MFCD17016068
CAS Number: 1618-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR