3-Carbamoyl-5-chlorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-Carbamoyl-5-chlorophenyl)boronic acid

A479873-100MG A479873-1G A479873-250MG
76877-826EA 36.08 USD
76877-826 76877-828 76877-830
3-Carbamoyl-5-chlorophenylboronic acid 95%
3-Carbamoyl-5-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClNO₃
MW: 199.4000 g/mol
MDL Number: MFCD09800858
CAS Number: 957120-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR