Methyl-1-methyl-1H-indole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-indole-5-carboxylate

A479554-1G A479554-250MG A479554-5G
76877-820EA 42.44 USD
76877-820 76877-822 76877-824
Methyl-1-methyl-1H-indole-5-carboxylate 97%
Methyl-1-methyl-1H-indole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.22 g/mol
MDL Number: MFCD11977801
CAS Number: 128742-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR