5-Bromo-N-methyl-2-nitroaniline 98%

Supplier: Ambeed
Danger

A477877-1G A477877-250MG
76877-800EA 31.83 USD
76877-800 76877-802
5-Bromo-N-methyl-2-nitroaniline 98%
5-Bromo-N-methyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD09862513
CAS Number: 302800-13-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR