3-(tert-Butyl)-5-iodo-1H-pyrazole 98%

Supplier: Ambeed
Warning

A476668-100MG A476668-1G A476668-250MG A476668-5G
76877-792EA 9 USD
76877-792 76877-794 76877-796 76877-798
3-(tert-Butyl)-5-iodo-1H-pyrazole 98%
5-tert-Butyl-3-iodo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁IN₂
MDL Number: MFCD13195724
CAS Number: 1218791-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR