Silver bis(trifluoromethanesulfonyl)imide 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
Silver Triflimide, Silver bis(trifluoromethane sulphonimide)
A475954-1G
A475954-250MG
A475954-25G
A475954-5G
76877-784EA
12
USD
76877-784
76877-786
76877-788
76877-790
Silver bis(trifluoromethanesulfonyl)imide 96%
Silver bis(trifluoromethanesulfonyl)imide
Precaución: for Research Use Only
Formula:
C₂AgF₆NO₄S₂ |
MDL Number:
MFCD08457644 CAS Number: 189114-61-2 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...