4-Bromo-N-isopropylaniline 96%

Supplier: Ambeed
Warning

A475292-100MG A475292-1G A475292-250MG
76877-772EA 23.34 USD
76877-772 76877-774 76877-776
4-Bromo-N-isopropylaniline 96%
4-Bromo-N-isopropylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.1023 g/mol
MDL Number: MFCD08691756
CAS Number: 121086-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR