(2S)-2-Aminohept-6-enoic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Aminohept-6-enoic acid

A472812-100MG A472812-250MG
76877-760EA 115.64 USD
76877-760 76877-762
(2S)-2-Aminohept-6-enoic acid 95%
(2S)-2-Aminohept-6-enoic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.184 g/mol
MDL Number: MFCD06200653
CAS Number: 166734-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR