Boc-N-Me-Tyr·DCHA 98%

Supplier: Ambeed
Warning

Synonyms: Boc-N-Me-Tyr-OH.DCHA, Boc-N-Me-Tyr-OH·DCHA

A472085-1G A472085-250MG A472085-25G A472085-5G
76877-752EA 26 USD
76877-752 76877-754 76877-756 76877-758
Boc-N-Me-Tyr·DCHA 98%
Boc-N-Me-Tyr·DCHA

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅·C₁₂H₂₃N
MDL Number: MFCD00237569
CAS Number: 95105-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR