1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane 98%

Supplier: Ambeed
Warning

A471936-100MG A471936-1G A471936-250MG A471936-5G
76877-744EA 10.61 USD
76877-744 76877-746 76877-748 76877-750
1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane 98%
1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane

Precaución: for Research Use Only

Formula: C₁₆H₂₁O₃P
MW: 292.3099 g/mol
MDL Number: MFCD10567051
CAS Number: 97739-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR