(E)-3-(Furan-2-yl)acrylic acid 95%

Supplier: Ambeed
Warning

A471566-100G A471566-1G A471566-500G A471566-5G A471566-25G
76877-732EA 123 USD
76877-732 76877-734 76877-736 76877-738 77884-206
(E)-3-(Furan-2-yl)acrylic acid 95%
(E)-3-(Furan-2-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₃
MDL Number: MFCD11656544
CAS Number: 15690-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR