3-Bromo-1-propylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A470658-1G A470658-5G A470658-250MG
76877-722EA 42 USD
76877-722 76877-724 77884-205
3-Bromo-1-propylboronic acid pinacol ester 98%
3-Bromo-1-propylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₉H₁₈BBrO₂
MDL Number: MFCD10567053
CAS Number: 124215-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR