5,8-Diazaspiro[3.5]nonane dihydrochloride 97%

Supplier: Ambeed
Danger

A468568-100MG A468568-1G A468568-250MG
76877-688EA 28 USD
76877-688 76877-690 76877-692
5,8-Diazaspiro[3.5]nonane dihydrochloride 97%
5,8-Diazaspiro[3.5]nonane dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂
MDL Number: MFCD06796599
CAS Number: 1159824-37-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR