3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Nitro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid

A468418-1G A468418-250MG A468418-5G
76877-682EA 85 USD
76877-682 76877-684 76877-686
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BN₂O₅
MDL Number: MFCD07783873
CAS Number: 871332-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR