N-Ethyl 3-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: (3-(N-Ethylsulphamoyl)phenyl)boronic acid, 3-(Ethylsulphamoyl)benzeneboronic acid

A467929-100MG A467929-1G A467929-250MG
76877-676EA 36.08 USD
76877-676 76877-678 76877-680
N-Ethyl 3-boronobenzenesulfonamide 98%
N-Ethyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
MDL Number: MFCD07783855
CAS Number: 871329-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR