Methyl 2-(1H-imidazol-2-yl)acetate 95%
Supplier: Ambeed
|
Warning
|
A466633-100MG
A466633-1G
A466633-250MG
76877-662EA
60
USD
76877-662
76877-664
76877-666
Methyl 2-(1H-imidazol-2-yl)acetate 95%
Methyl 2-(1H-imidazol-2-yl)acetate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ |
MDL Number:
MFCD11844783 CAS Number: 1564-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...