cis-3-Carbomethoxycyclohexane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: cis-3-(Methoxycarbonyl)cyclohexanecarboxylic acid

A466559-100MG A466559-250MG
76877-658EA 91.24 USD
76877-658 76877-660
cis-3-Carbomethoxycyclohexane-1-carboxylic acid 98%
cis-3-Carbomethoxycyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
MDL Number: MFCD01311244
CAS Number: 733742-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR