1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A464592-100MG A464592-1G A464592-250MG
76877-650EA 85.94 USD
76877-650 76877-652 77673-188
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid 98%
1H-Pyrrolo[3,2-c]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
MDL Number: MFCD11845529
CAS Number: 1082040-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR