2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(2,2,2-Trifluoroethoxy)pyridin-3-yl)boronic acid

A463937-100MG A463937-1G A463937-250MG A463937-5G
76877-638EA 37.14 USD
76877-638 76877-640 76877-642 76877-644
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid 98%
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₃NO₃
MW: 220.9400 g/mol
MDL Number: MFCD07374887
CAS Number: 1218790-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR