(S)-2-(3-Chloro-2-hydroxypropyl)isoindoline-1,3-dione 95%

Supplier: Ambeed
Danger

A463115-100MG A463115-250MG
76877-618EA 20.16 USD
76877-618 76877-620
(S)-2-(3-Chloro-2-hydroxypropyl)isoindoline-1,3-dione 95%
(S)-2-(3-Chloro-2-hydroxypropyl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClNO₃
MW: 239.66 g/mol
MDL Number: MFCD18642725
CAS Number: 148857-42-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR