Benzyl 4-methylenepiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A461092-1G A461092-25G A461092-5G
76877-608EA 20.16 USD
76877-608 77568-908 77594-452
Benzyl 4-methylenepiperidine-1-carboxylate 98%
Benzyl 4-methylenepiperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₄H₁₇NO₂
MW: 231.29 g/mol
MDL Number: MFCD09834622
CAS Number: 138163-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR