5-Bromo-2,4-difluorophenol 96%

Supplier: Ambeed
Warning

A459955-1G A459955-250MG A459955-5G
76877-600EA 55 USD
76877-600 76877-602 76877-604
5-Bromo-2,4-difluorophenol 96%
5-Bromo-2,4-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MDL Number: MFCD20231458
CAS Number: 355423-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR