4-Fluoro-1H-indole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A458375-100MG A458375-1G A458375-250MG
76877-582EA 41.38 USD
76877-582 76877-584 76877-586
4-Fluoro-1H-indole-3-carboxylic acid 96%
4-Fluoro-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.14 g/mol
MDL Number: MFCD09954941
CAS Number: 23077-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR