Tetrahydro-2H-thiopyran-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

A457602-100MG A457602-1G A457602-250MG A457602-5G
76877-574EA 16.98 USD
76877-574 76877-576 76877-578 76877-580
Tetrahydro-2H-thiopyran-4-amine hydrochloride 95%
Tetrahydro-2H-thiopyran-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNS
MDL Number: MFCD09701226
CAS Number: 233763-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR