5-Benzylcarbamoyl-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A452909-100MG A452909-1G A452909-250MG
76877-534EA 34 USD
76877-534 76877-536 76877-538
5-Benzylcarbamoyl-2-fluorophenylboronic acid 97%
5-Benzylcarbamoyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₃
MDL Number: MFCD08436031
CAS Number: 874289-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR