tert-Butyl 3-(methylamino)propionate 95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 3-(methylamino)propanoate

A452148-250MG A452148-1G A452148-5G A452148-100MG
76877-522EA 53 USD
76877-522 77594-412 77673-128 77884-162
tert-Butyl 3-(methylamino)propionate 95%
tert-Butyl 3-(methylamino)propionate

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂
MW: 159.226 g/mol
MDL Number: MFCD08275841
CAS Number: 143707-72-6
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR