trans-4-(Hydroxymethyl)cyclohexanol 98%
Supplier: Ambeed
|
Warning
|
A450938-100MG
A450938-250MG
A450938-5G
76877-502EA
14
USD
76877-502
76877-504
77884-156
trans-4-(Hydroxymethyl)cyclohexanol 98%
trans-4-(Hydroxymethyl)cyclohexanol
Precaución: for Research Use Only
Formula:
C₇H₁₄O₂ |
MDL Number:
MFCD11036248 CAS Number: 3685-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...