3-Fluor-4-(propylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (3-Fluoro-4-(propylcarbamoyl)phenyl)boronic acid

A448718-1G A448718-250MG A448718-5G
76877-474EA 128.37 USD
76877-474 76877-476 76877-478
3-Fluor-4-(propylcarbamoyl)phenylboronic acid 98%
3-Fluor-4-(propylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₃
MW: 225.0300 g/mol
MDL Number: MFCD09258737
CAS Number: 874289-15-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR