2-(Methylthio)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A448078-25G A448078-5G
76877-472EA 187 USD
76877-472 77551-348
2-(Methylthio)ethanamine hydrochloride 95%
2-(Methylthio)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₀ClNS
MDL Number: MFCD02755761
CAS Number: 6950-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR