(2-Fluorobenzyl)hydrazine hydrochloride ≥98%

Supplier: Ambeed
Warning

A445936-1G A445936-25G A445936-5G
76877-452EA 14 USD
76877-452 76877-454 76877-456
(2-Fluorobenzyl)hydrazine hydrochloride ≥98%
(2-Fluorobenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀ClFN₂
MDL Number: MFCD12403346
CAS Number: 1216246-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR