1-(((tert-Butoxycarbonyl)amino)methyl)cyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A445615-100MG A445615-1G A445615-250MG
76877-446EA 46.68 USD
76877-446 76877-448 76877-450
1-(((tert-Butoxycarbonyl)amino)methyl)cyclopentanecarboxylic acid 95%
1-(((tert-Butoxycarbonyl)amino)methyl)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.29 g/mol
MDL Number: MFCD09264201
CAS Number: 204514-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR