8-Fluoroisoquinoline 98%

Supplier: Ambeed
Warning

A445046-1G A445046-250MG A445046-5G
76877-440EA 143.23 USD
76877-440 76877-442 76877-444
8-Fluoroisoquinoline 98%
8-Fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆FN
MW: 147.1500 g/mol
MDL Number: MFCD09878369
CAS Number: 1075-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR