4-Bromo-5-methylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A444899-100MG A444899-1G A444899-250MG A444899-5G
76877-424EA 71.09 USD
76877-424 76877-426 76877-428 77551-340
4-Bromo-5-methylpyridin-2-amine 95%
4-Bromo-5-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD11100640
CAS Number: 1033203-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR