3-Bromo-4-fluoro-5-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A443687-10G A443687-1G A443687-250MG A443687-5G
76877-408EA 100 USD
76877-408 76877-410 76877-412 76877-414
3-Bromo-4-fluoro-5-nitrobenzoic acid 97%
3-Bromo-4-fluoro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₄
MDL Number: MFCD18914494
CAS Number: 1290117-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR