(2R,5S)-rel-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A442830-1G A442830-5G
76877-404EA 54 USD
76877-404 77886-180
(2R,5S)-rel-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate 98%
(2R,5S)-rel-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD16036945
CAS Number: 194032-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR