4-(Trifluoromethoxy)benzenesulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
A442456-10G
A442456-1G
A442456-5G
A442456-25G
76877-392EA
19
USD
76877-392
76877-394
76877-396
77551-332
4-(Trifluoromethoxy)benzenesulfonyl chloride 98%
4-(Trifluoromethoxy)benzenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₇H₄ClF₃O₃S |
MDL Number:
MFCD00042408 CAS Number: 94108-56-2 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...