6-Chloropyridin-2-ol 95%

Supplier: Ambeed
Warning

A441520-100G A441520-25G A441520-5G
76877-386EA 95.49 USD
76877-386 76877-388 76877-390
6-Chloropyridin-2-ol 95%
6-Chloropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₄ClNO
MW: 129.54 g/mol
MDL Number: MFCD00006278
CAS Number: 73018-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR