6-Amino-4-hydroxy-2-naphthalenesulfonic acid 97% (by HPLC, titration analysis) (contains < 10% H₂O)

Supplier: Ambeed
Warning

Synonyms: gamma acid, 7-Amino-1-naphthol-3-sulphonic acid, 2-Amino-8-naphthol-6-sulphonic acid

A440526-1000G A440526-100G A440526-25G
76877-372EA 82.76 USD
76877-372 76877-374 76877-376
6-Amino-4-hydroxy-2-naphthalenesulfonic acid 97% (by HPLC, titration analysis) (contains < 10% H₂O)
6-Amino-4-hydroxy-2-naphthalenesulphonic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
MDL Number: MFCD00003973
CAS Number: 90-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR